Bootie cabinets
Bootie Cabinet 48, 683×400×2100 mm
Catalog # 134-0477
Supplier: Palbam
Gå til:
Product Details & Documents
The modular bootie cabinets come in a variety of shelf lengths.
- Supplied with leveler legs and foot covers
- This symetric product can be used from either side
- Adhesive labelling numbers supplied
Construction: Stainless steel 304L with electropolished finish
: Typically available from stock for direct shipment from the supplier.
Documents
Palbam Class - Catalog 2018
Product Literature
Specifikationer
- Description:Bootie Cabinet 48
- W×D×H:683×400×2100 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



