Bootie cabinets
Bootie Cabinet 40, 583×400×2100 mm
Catalog # 134-0476
Supplier: Palbam
Jump to:
Product Details & Documents
The modular bootie cabinets come in a variety of shelf lengths.
- Supplied with leveler legs and foot covers
- This symetric product can be used from either side
- Adhesive labelling numbers supplied
Construction: Stainless steel 304L with electropolished finish
Delivery information: Typically available from stock for direct shipment from the supplier.
Documents
Palbam Class - Catalog 2018
Product Literature
Specifications
- Description:Bootie Cabinet 40
- W×D×H:583×400×2100 mm



