Order Entry
Denmark
Orders LinkContactUsLinkComponent
Bootie cabinets

Bootie cabinets

Bootie Cabinet 32, 483×400×2100 mm
  134-0475
 :   Palbam
Bootie cabinets
  134-0475
 :  Palbam
Bootie cabinets
Bootie cabinets

Bootie Cabinet 32, 483×400×2100 mm

 
39.080,00 kr
   
1 * 1 Items
 

The modular bootie cabinets come in a variety of shelf lengths.

  • Supplied with leveler legs and foot covers
  • This symetric product can be used from either side
  • Adhesive labelling numbers supplied

Construction: Stainless steel 304L with electropolished finish

Delivery information: Typically available from stock for direct shipment from the supplier.

 
  • Description:
    Bootie Cabinet 32
  • W×D×H:
    483×400×2100 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.