Order Entry
Denmark
ContactUsLinkComponent
Flask, volumetric, 50ml, NS 12/21, trapezoidal shape, blue graduated, poly stopper, class A 1 * 2 items
Catalog # HIRS2960175_S
Supplier:  HIRSCHMANN
undefined
Flask, volumetric, 50ml, NS 12/21, trapezoidal shape, blue graduated, poly stopper, class A 1 * 2 items
Catalog # HIRS2960175_S
Supplier:  HIRSCHMANN
Frequently Bought Together