Order Entry
Denmark
ContactUsLinkComponent
Flask volumetric, capacity 100ml, with rim, DURAN, class A, tolerance 0.1ml, height 170mm, graduation color blue, conformity certifi ed. 1 * 1 items
Catalog # HIRS2800180_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 100ml, with rim, DURAN, class A, tolerance 0.1ml, height 170mm, graduation color blue, conformity certifi ed. 1 * 1 items
Catalog # HIRS2800180_S
Supplier:  HIRSCHMANN
Frequently Bought Together