Order Entry
Denmark
ContactUsLinkComponent
Flask volumetric, capacity 5ml, with rim, DURAN, class A, tolerance 0.025ml, height 70mm, graduation color blue, conformity certifie d. 1 * 1 items
Catalog # HIRS2800152_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 5ml, with rim, DURAN, class A, tolerance 0.025ml, height 70mm, graduation color blue, conformity certifie d. 1 * 1 items
Catalog # HIRS2800152_S
Supplier:  HIRSCHMANN
Frequently Bought Together