Order Entry
Denmark
ContactUsLinkComponent
Flask volumetric, capacity 50ml, class A, amber graduated, glass stoppered, tolerance 0.08ml, height 140mm, NS-ground 14/23, graduat ion color white, conformity certified. 1 * 1 items
  HIRS2640278_S
undefined
Flask volumetric, capacity 50ml, class A, amber graduated, glass stoppered, tolerance 0.08ml, height 140mm, NS-ground 14/23, graduat ion color white, conformity certified. 1 * 1 items
  HIRS2640278_S
Frequently Bought Together