Order Entry
Denmark
ContactUsLinkComponent
Flask volumetric, capacity 250ml, class A, amber graduated, polystoppered, tolerance 0.15ml, height 220mm, NS-ground 14/23, graduati on color white, conformity certified. 1 * 2 items
Catalog # HIRS2640186_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 250ml, class A, amber graduated, polystoppered, tolerance 0.15ml, height 220mm, NS-ground 14/23, graduati on color white, conformity certified. 1 * 2 items
Catalog # HIRS2640186_S
Supplier:  HIRSCHMANN
Frequently Bought Together