Flask volumetric, capacity 50ml, class A, amber graduated, polystoppered, tolerance 0.08ml, height 140mm, NS-ground 14/23, draduatio n color white, conformity certified. 1 * 2 items
Catalog # HIRS2640178_S
Supplier: HIRSCHMANN
Frequently Bought Together