Order Entry
Denmark
ContactUsLinkComponent
Flask volumetric, capacity 10ml, class A, amber graduated, poly stoppered, tolerance 0.025ml, height 90mm, NS-ground 7/16, graduatio n color white, conformity certified. 1 * 2 items
Catalog # HIRS2640160_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 10ml, class A, amber graduated, poly stoppered, tolerance 0.025ml, height 90mm, NS-ground 7/16, graduatio n color white, conformity certified. 1 * 2 items
Catalog # HIRS2640160_S
Supplier:  HIRSCHMANN
Frequently Bought Together