Order Entry
Denmark
ContactUsLinkComponent
Flask volumetric, capacity 5ml, class A, brown dyed continuously, tolerance 0.04ml, height 70mm, NS-ground 10/19, graduation color w hite, poly stoppered, conformity certified. 1 * 2 items
Catalog # HIRS2640153_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 5ml, class A, brown dyed continuously, tolerance 0.04ml, height 70mm, NS-ground 10/19, graduation color w hite, poly stoppered, conformity certified. 1 * 2 items
Catalog # HIRS2640153_S
Supplier:  HIRSCHMANN
Frequently Bought Together