Order Entry
Denmark
ContactUsLinkComponent
Pipette volumetric, capacity 50ml, class AS, blue graduation, tolerance 0.05ml, total length +-10mm:540mmconformity certified. 1 * 1 items
Catalog # HIRS1341175_S
Supplier:  HIRSCHMANN
undefined
Pipette volumetric, capacity 50ml, class AS, blue graduation, tolerance 0.05ml, total length +-10mm:540mmconformity certified. 1 * 1 items
Catalog # HIRS1341175_S
Supplier:  HIRSCHMANN
Frequently Bought Together