Order Entry
Denmark
ContactUsLinkComponent
Pipette volumetric, capacity 1ml, class AS, blue graduation, tolerance 0.007ml, total length +-10mm:300mm, conformity certified. 1 * 1 items
Catalog # HIRS1341133_S
Supplier:  HIRSCHMANN
undefined
Pipette volumetric, capacity 1ml, class AS, blue graduation, tolerance 0.007ml, total length +-10mm:300mm, conformity certified. 1 * 1 items
Catalog # HIRS1341133_S
Supplier:  HIRSCHMANN
Frequently Bought Together