Order Entry
Denmark
Orders LinkContactUsLinkComponent
Aluminium inserts for ice baths
Aluminium inserts for ice baths
Aluminium insert for 2×12 conical reaction vials (1,5 ml), for TOP ice bath (large)
Catalog # 216-6901
Aluminium inserts for ice baths
Catalog # 216-6901
Aluminium inserts for ice baths
Aluminium inserts for ice baths
Aluminium insert for 2×12 conical reaction vials (1,5 ml), for TOP ice bath (large)
Price
1.680,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
The inserts are simply mounted in the special groove.
Specifications
  • Description:
    Aluminium insert for 2×12 conical reaction vials (1,5 ml), for TOP ice bath (large)
  • Seq:
    02