Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic

Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic

R200 clamp for fastening the T 10 basic to the stand, T 10 basic
Catalog # 442-0313
Supplier:   IKA
Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic
Catalog # 442-0313
Supplier:  IKA
Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic
Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic

R200 clamp for fastening the T 10 basic to the stand, T 10 basic

Price
364,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    R200 clamp for fastening the T 10 basic to the stand
  • Seq:
    106
  • For:
    T 10 basic
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Strap clamp for securing vessels, stand Ø 25 - 36 mm, vessel Ø 40 - 300 mm
    Strap clamp for securing vessels, stand Ø 25 - 36 mm, vessel Ø 40 - 300 mm
    431-2944
    Log in for availability
    1 * 1 Items - 3.570,00 kr

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.