Order Entry
Denmark
Orders LinkContactUsLinkComponent
Colony counters
Colony counters
Colony counter manual with magnifying glass ø 100 mm with support
Catalog # 710-0107
Colony counters
Catalog # 710-0107
Supplier:  
Colony counters
Colony counters
Colony counter manual with magnifying glass ø 100 mm with support
Discontinued
Product Details & Documents
Choice of models with a fibre marker with a built-in switch and /or a metal-tipped pen that are attached to the base. The marker supplied with the counter allows marked cells to be added to the screen one by one.
  • Ø 100 or 120 mm biconvex lamp
  • X2 magnification
220 V voltage, 50 Hz, 0,2 A.
Specifications
  • Description:
    Colony counter manual with magnifying glass ø 100 mm with support
  • Seq:
    01