Order Entry
Denmark
Orders LinkContactUsLinkComponent
Rods, AR-Glas®

Rods, AR-Glas®

6±0,15 mm, 1500 mm
Catalog # SCOR1028430
Supplier:   Schott AG
Rods, AR-Glas®
Catalog # SCOR1028430
Supplier:  Schott AG
Rods, AR-Glas®

6±0,15 mm, 1500 mm

Price
1.440,00 kr
Log in for availability
1 * 15 kg
Product Details & Documents

Made of AR®-Glas.

  • Soft glass is acid-resistant
  • Has high thermal expansion for pressure melting and for fusing with platinum or dumet and other alloys
Specifications
  • Ext.Ø:
    6±0,15 mm
  • Length:
    1500 mm
  • Seq:
    04