Forceps
Super fine point, straight, 115 mm
Catalog # 241-0332
Supplier: EXCELTA
Jump to:
Product Details & Documents
Stainless steel, antimagnetic.
- Economical special forceps for precision applications
- Corrosion-resistant
Ideal when working with microelectronic assemblies and in the electronics industry.
Specifications
- Description:Super fine point, straight
- Length:115 mm
- Seq:03