Order Entry
Denmark
Orders LinkContactUsLinkComponent
2-Thiobarbituric acid ≥98%

2-Thiobarbituric acid ≥98%

Glass bottle for solids, 25 g
Catalog # A12681.14
2-Thiobarbituric acid ≥98%
Catalog # A12681.14
CAS Number:  504-17-6
2-Thiobarbituric acid ≥98%
2-Thiobarbituric acid ≥98%

Glass bottle for solids, 25 g

CAS Number:  504-17-6
Price
612,00 kr
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 504-17-6 Molecular Formula: C4H4N2O2S Formula Weight: 144.15 pH: 1.8 Melting Point: 300° Beilstein Reference: 120663 MDL No.: MFCD00006674

Synonyms:  4,6-Dihydroxy-2-mercaptopyrimidine
  • CAS Number:504-17-6
  • Formula:C₄H₄N₂O₂S
  • MDL Number:MFCD00006674
  • Molecular Weight:144.15 g/mol
  • Boiling Pt:419 °C (1013 hPa)
  • Melting Pt:235 °C
  • Density:1.661 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-985-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.