Modules, Immuno™ LockWell™ Fluoronunc™/Luminunc™
Black modules, framed, C8, MaxiSorp surface, Black, 96
Catalog # 738-0750
Supplier: Thermo Fisher Scientific
Modules, Immuno™ LockWell™ Fluoronunc™/Luminunc™
Catalog # 738-0750
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PS, 96 wells per frame. For fluorescence/luminescence-based immunoassays and binding assays, these modules have been designed using the standard 96 MicroWell™ LockWell™ format. Opaque white modules have a reflective surface and offer low cross talk and minimal background. Opaque black modules with light absorbing surface offer minimum cross talk and back scattered light.
- Each well locks in the frame of the same height to prevent jamming in machines
- Easy to handle
- Uniform washing and reading
- Certified binding homogeneity for all four surfaces
- MaxiSorp™ surface has a high affinity to molecules with mixed hydrophilic/hydrophobic domains; PolySorp™ surface has a high affinity for hydrophobic molecules
Total volume per well: 350 µl
Specifications
- No. of wells:96
- Colour:Black
- Description:Black modules, framed, C8, MaxiSorp surface
- Cat. No.:738-0750
- Environmental certification:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact