pH buffer solutions, referenced at 20 °C, Fluka™
10,00, ±0.05, Borate, 20 °C, Plastic bottle, 1×500 ml, 500 mL
Supplier: Honeywell Chemicals
pH buffer solutions, referenced at 20 °C, Fluka™
Catalog # 33649-500ML
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Documents
Specifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Borate
- Pk:500 mL
- Reference temperature:20 °C
- pH accuracy:±0.05
- Pk Info:1×500 ml
- pH value:10,00



