Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spatulas, round, bent

Spatulas, round, bent

185 mm, 10×5 mm
Catalog # BOCH3201
Supplier:   BOCHEM
Spatulas, round, bent
Catalog # BOCH3201
Supplier:  BOCHEM
Spatulas, round, bent
Spatulas, round, bent

185 mm, 10×5 mm

Price
19,90 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Stainless steel ¹⁸/₁₀, anti-magnetic. These spatulas with one rounded end can be used for a variety of applications.

  • Excellent heat resistance, autoclavable
  • Good resistance to chemicals
  • Easy and quick cleaning

Documents

Specifications
  • Length:
    185 mm
  • Seq:
    04
  • Spoon size:
    10×5 mm