Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spatulas, round, bent

Spatulas, round, bent

130 mm, 10×5 mm
Catalog # BOCH32005
Supplier:   BOCHEM
Spatulas, round, bent
Catalog # BOCH32005
Supplier:  BOCHEM
Spatulas, round, bent
Spatulas, round, bent

130 mm, 10×5 mm

Price
92,50 kr
Log in for availability
1 * 5 Items
Product Details & Documents

Stainless steel ¹⁸/₁₀, anti-magnetic. These spatulas with one rounded end can be used for a variety of applications.

  • Excellent heat resistance, autoclavable
  • Good resistance to chemicals
  • Easy and quick cleaning

Documents

Specifications
  • Length:
    130 mm
  • Seq:
    02
  • Spoon size:
    10×5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.