Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cytoplasmic membrane labelling kits, CellBrite™

Cytoplasmic membrane labelling kits, CellBrite™

CellBrite™ Orange, 549/565 nm, 1 mL
Catalog # 30022.
Supplier:   Biotium
Cytoplasmic membrane labelling kits, CellBrite™
Catalog # 30022.
Supplier:  Biotium
CAS Number:  
Cytoplasmic membrane labelling kits, CellBrite™
Cytoplasmic membrane labelling kits, CellBrite™

CellBrite™ Orange, 549/565 nm, 1 mL

CAS Number:  
Price
1.770,00 kr
Log in for availability
1 * 1 mL
Product Details & Documents

CellBrite kits are designed to label cell cytoplasmic membranes with a fluorescent membrane dye pre-dissolved as an optimised staining solution. By combining CellBrite membrane stains, it is possible to label multiple cell populations with distinctive colours, a capability useful for the studies of cell-cell interactions.

  • Direct addition of the dye to normal culture media followed by a brief incubation period
  • Highly lipophilic dye ensuring no dye transfer between cells, resulting in stable labelling
  • Bright, photostable, and non toxic

The labelling is stable and non toxic, suitable for long-term tracking of cells.

Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    CellBrite™ Orange
  • Abs/Em:
    549/565 nm

Specifications

Complementary Products

Product Information

  • Catalog #
    Description
    Model
    Resolution
    Serum format
    Sterility
    Recommended working volume
    Colour
    Packed
    Sterile
    Working volume
    Availability
    Price
  • MLDVMICRO4
    High content imaging system
    ImageXpress Micro 4
    25 - 100 nm
    Log in for availability
    1 * 1 Items - kr0.00

  • 735-0364
    High-content micro-confocal imaging system
    MICROCONFOCAL
    01
    Log in for availability
    1 * 1 Items - kr0.00

  • S1810-500
    Foetal bovine serum, South American origin
    Frozen
    Sterile filtered
    Log in for availability
    1 * 500 mL - kr2,400.00

  • 734-2097
    200 µl
    96-MicroWell™ plate, flat bottomed wells, with lid
    06
    Log in for availability
    1 * 50 Items - kr1,630.00

  • CORN3904BC
    20/bag
    96-well microplate, clear flat bottom, tissue culture treated, with lid
    +
    21
    360 µl
    Black
    Log in for availability
    1 * 100 Items - kr9,690.00

  • 735-0523
    Microlite™ 1+ strip, 1×12 per strip
    Log in for availability
    1 * 320 Items - kr1,710.00

  • 734-2327
    1/bag
    0,075 - 0,20 ml
    96-well plates, flat bottom, TC-treated
    +
    17
    Clear
    Log in for availability
    1 * 100 Items - kr1,940.00