Order Entry
Denmark
Orders LinkContactUsLinkComponent
Baicalein

Baicalein

Glass bottle for solids, 50 mg
Catalog # CAYM70610-50
Supplier:   Cayman Chemical
Baicalein
Catalog # CAYM70610-50
Supplier:  Cayman Chemical
CAS Number:  491-67-8
Baicalein

Glass bottle for solids, 50 mg

CAS Number:  491-67-8
Price
224,00 kr
Log in for availability
1 * 50 mg
Product Details & Documents
Synonyms:  5,6,7-Trihydroxyflavone, 5,6,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 5,6,7-Trihydroxy-2-phenylchromen-4-one
  • CAS Number:491-67-8
  • Formula:C₁₅H₁₀O₅
  • MDL Number:MFCD00017459
  • Molecular Weight:270.24 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg

Specifications