Disposable base moulds
Base mould, 15×15×5 mm
Catalog # 720-0821
Supplier: Simport Scientific
Jump to:
Product Details & Documents
These disposable PVC base moulds offer ease and convenience of operation. They can be discarded after use, but are also strong enough to be reused.
- Excellent thermal exchange
- Smooth interior
- Rounded corners
They are available in the same variety of sizes as metal moulds and can be used with the same styles or types of cassettes and embedding rings.
: Each case contains two dispenser boxes of 500 base moulds.
Specifications
- Description:Base mould
- Dimensions:15×15×5 mm
- Seq:02