Order Entry
Denmark
Orders LinkContactUsLinkComponent
PCR plates
PCR plates
24, Non-skirted, Red
Catalog # 211-3398
Supplier:   Simport Scientific
PCR plates
Catalog # 211-3398
Supplier:  Simport Scientific
PCR plates
PCR plates
24, Non-skirted, Red
Price
564,00 kr
Log in for availability
1 * 50 Items
Product Details & Documents
PP. Thin walled PCR plates compatible with 0,2 ml thermal cyclers. The insides of the tubes are smooth and have an inert surface on which enzymes and nucleic acids do not bind.
  • Alphanumeric grid for better identification
  • Flexible plate for better fitting of tubes in the thermal block
  • Larger plates are easily cut into sections of 24, 32 or 48 tubes, but, for more convenience, pre-cut plates are also available with formats of 48 tubes (6×8) and 24 tubes (3×8)
  • The bottom right corner of the plate is cut away to facilitate orientation
  • Certified free of RNase, DNase and DNA
Packaging: Packed in tamperproof, resealable bags of 10 plates.
Specifications
  • No. of wells:
    24
  • Skirt:
    Non-skirted
  • Well colour:
    Red