Tissue processing/embedding cassettes, Histosette® II, M492
Pale orange
720-1528
Acetal polymer.
- Resistant to the chemical action of histological solvents
- Flow-through slots maximise fluid exchange
- Integral lid
- Anterior writing area at 45° angle to enable use with leading cassette labelling instruments
- Colour:Pale orange
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



