Tissue processing/embedding cassettes, Histosette® II, M492
Turquoise
Catalog # 720-0908
Supplier: Simport Scientific
Tissue processing/embedding cassettes, Histosette® II, M492
Catalog # 720-0908
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Acetal polymer.
- Resistant to the chemical action of histological solvents
- Flow-through slots maximise fluid exchange
- Integral lid
- Anterior writing area at 45° angle to enable use with leading cassette labelling instruments
Specifications
- Colour:Turquoise
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



