1-Bromo-2-chloroethane
Glass ampoule, 1 Ampoul
Katalog # REST30469-3
Leverandør: Restek
Gå til:
Product Details & Documents
Synonymer: Ethylene chlorobromide
- CAS nummer:107-04-0
- Formel:BrH₂CCH₂Cl
- MDL nummer:MFCD00000962
- Molecular Weight:143.41 g/mol
- Kogende Pt:107 °C (1013 hPa)
- Smeltende Pt:–17 °C
- Tæthed:1.7392 g/cm³ (20 °C)
- Opbevaringstemperatur:Ambient
- EINECS:203-456-0
- FN:2810
- ADR:6.1,III
Specifikationer
Specifikationer
- Pack type:Glass ampoule
- Pk:1 Ampoul
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



