Order Entry
Denmark
Orders LinkContactUsLinkComponent
Replacement Cables
Replacement Cables
Replacement cable. Connect an Agilent 5890 GC to a non-Agilent integrator or standard strip chart recorder, 1,8 m
Catalog # REST20652
Supplier:   Restek
Replacement Cables
Catalog # REST20652
Supplier:  Restek
Replacement Cables
Replacement Cables
Replacement cable. Connect an Agilent 5890 GC to a non-Agilent integrator or standard strip chart recorder, 1,8 m
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
These are replacement cables for Agilent GCs, integrators and autosamplers.
Delivery information: Instructions and writing diagrams included.
Specifications
  • Description:
    Replacement cable. Connect an Agilent 5890 GC to a non-Agilent integrator or standard strip chart recorder
  • Length:
    1,8 m
  • Seq:
    01