Solid phase extraction cartridges, Resprep®, method specific
EPA method 548.1, for extraction of endothall from aqueous samples, PP, 6 ml, -
Catalog # REST26063
Supplier: Restek
Jump to:
Product Details & Documents
Cartridges have been specifically designed to provide consistent and reproducible results for the method or application stated.
Specifications
- Capacity:6 ml
- Description:EPA method 548.1, for extraction of endothall from aqueous samples
- Seq:03
- Bed weight:-
- Column hardware:PP
Anbefalinger