Order Entry
Denmark
Orders LinkContactUsLinkComponent
Precision sampling valves for vials, Mininert®
Precision sampling valves for vials, Mininert®
Mininert® precision sampling valves, for screw-thread vials, 18 mm/400 thread
Catalog # REST24902
Supplier:   Restek
Precision sampling valves for vials, Mininert®
Catalog # REST24902
Supplier:  Restek
Precision sampling valves for vials, Mininert®
Precision sampling valves for vials, Mininert®
Mininert® precision sampling valves, for screw-thread vials, 18 mm/400 thread
Price
1.730,00 kr
Log in for availability
1 * 12 Items
Product Details & Documents
Mininert® valves are very convenient for repetitive sampling and limit content exposure to the silicon septum. The crimp-top valve for 20 mm ID glassware slides into the neck of the vial.
  • Available for screw cap and crimp top vials
  • Threaded flange used to secure tight fit
Specifications
  • Description:
    Mininert® precision sampling valves, for screw-thread vials, 18 mm/400 thread
  • Seq:
    03
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Replacement septa
    Replacement septa
    REST24906
    Log in for availability
    1 * 50 Items - 485,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Septum insertion tool
    Septum insertion tool
    REST24907
    Log in for availability
    1 * 1 Items - 441,00 kr