Solid phase extraction cartridges, Resprep®
SPE silica cartridges, with PTFE frits, Cartridge, 3 ml, 500 mg
REST24036
: Restek
These cartridges are manufactured using high density polypropylene and have polyethylene frits unless otherwise noted. Hydrophilic (polar) adsorbents used to extract hydrophilic analytes from nonpolar matrices, such as organic solvents (e.g., polar contaminants from sample extracts).
Cartridges may be processed by any one or all of these techniques: positive pressure, sidearm flask, centrifuge, or vacuum manifold.
- Column Format:Cartridge
- Capacity:3 ml
- Description:SPE silica cartridges, with PTFE frits
- Seq:05
- Bed weight:500 mg