Order Entry
Denmark
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Resprep®
SPE Florisil cartridges, Cartridge, 6 ml, 1000 mg
  REST24034
 :   Restek
Solid phase extraction cartridges, Resprep®
  REST24034
 :  Restek
Solid phase extraction cartridges, Resprep®
Solid phase extraction cartridges, Resprep®
SPE Florisil cartridges, Cartridge, 6 ml, 1000 mg
1.990,00 kr
1 * 30 Items
These cartridges are manufactured using high density polypropylene and have polyethylene frits unless otherwise noted. Hydrophilic (polar) adsorbents used to extract hydrophilic analytes from nonpolar matrices, such as organic solvents (e.g., polar contaminants from sample extracts).
Cartridges may be processed by any one or all of these techniques: positive pressure, sidearm flask, centrifuge, or vacuum manifold.
  • Column Format:
    Cartridge
  • Capacity:
    6 ml
  • Description:
    SPE Florisil cartridges
  • Seq:
    03
  • Bed weight:
    1000 mg