Order Entry
Denmark
Orders LinkContactUsLinkComponent
Rethreading tool
Rethreading tool, for 7/16" compression fitting (Bruker/Varian injection ports)
  REST23019
 :   Restek
Rethreading tool
  REST23019
 :  Restek
Rethreading tool
Rethreading tool
Rethreading tool, for 7/16" compression fitting (Bruker/Varian injection ports)
1 * 1 Items
These rethreading tools are suitable for repairing worn or damaged threads that come as a result of consistent use and temperature changes.
  • Multiple uses (injection ports, fittings, etc.)
  • Built-in guide prevents cross-threading
Repeated installation, removal and temperature changes cause wear and damage to threads on GC parts. This can cause a poor seal, and oxygen can enter the system, compromising analytical results and possibly destroying expensive analytical columns.
  • Description:
    Rethreading tool, for 7/16" compression fitting (Bruker/Varian injection ports)
  • Seq:
    04