Order Entry
Denmark
Orders LinkContactUsLinkComponent
Injector adapters
Injector adapters
Injector adapter for PerkinElmer CAP injector, stainless steel, for PE-style capillary nuts, PE-style capillary nuts
Katalog # REST22320
Leverandør:   Restek
Injector adapters
Katalog # REST22320
Leverandør:  Restek
Injector adapters
Injector adapters
Injector adapter for PerkinElmer CAP injector, stainless steel, for PE-style capillary nuts, PE-style capillary nuts
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These injector adapters made of stainless steel are ideal for PerkinElmer CAP injector.
Siltek®-treated version available for increased inertness.
Specifikationer
  • For:
    PE-style capillary nuts
  • Description:
    Injector adapter for PerkinElmer CAP injector, stainless steel, for PE-style capillary nuts
  • Seq:
    02
Anbefalinger