Order Entry
Denmark
Orders LinkContactUsLinkComponent
Injector adapters
Injector adapters
Injector adapter for PerkinElmer CAP injector, Siltek®-treated, for 1/16" compression-style nuts, 1/16" compression-style nuts
Catalog # REST22319
Supplier:   Restek
Injector adapters
Catalog # REST22319
Supplier:  Restek
Injector adapters
Injector adapters
Injector adapter for PerkinElmer CAP injector, Siltek®-treated, for 1/16" compression-style nuts, 1/16" compression-style nuts
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
These injector adapters made of stainless steel are ideal for PerkinElmer CAP injector.
Siltek®-treated version available for increased inertness.
Specifications
  • For:
    1/16" compression-style nuts
  • Description:
    Injector adapter for PerkinElmer CAP injector, Siltek®-treated, for 1/16" compression-style nuts
  • Seq:
    03