Order Entry
Denmark
Orders LinkContactUsLinkComponent
Rethreading tool
Rethreading tool, for 1/4" Bruker/Varian-style capillary column fittings
Catalog # REST21893
Supplier:   Restek
Rethreading tool
Catalog # REST21893
Supplier:  Restek
Rethreading tool
Rethreading tool
Rethreading tool, for 1/4" Bruker/Varian-style capillary column fittings
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
These rethreading tools are suitable for repairing worn or damaged threads that come as a result of consistent use and temperature changes.
  • Multiple uses (injection ports, fittings, etc.)
  • Built-in guide prevents cross-threading
Repeated installation, removal and temperature changes cause wear and damage to threads on GC parts. This can cause a poor seal, and oxygen can enter the system, compromising analytical results and possibly destroying expensive analytical columns.
Specifications
  • Description:
    Rethreading tool, for 1/4" Bruker/Varian-style capillary column fittings
  • Seq:
    05