Capillary installation gauges
Capillary installation gauge for 1/16" fittings (1/16" ferrules)
Catalog # REST21399
Supplier: Restek
Jump to:
Product Details & Documents
These capillary installation gauges for Agilent GCs are made from high-quality stainless steel.
- Prevents crushed column ends
- Seats graphite ferrules onto column for consistent installations
Specifications
- Description:Capillary installation gauge for 1/16" fittings (1/16" ferrules)
- Seq:02