Order Entry
Denmark
Orders LinkContactUsLinkComponent
Fingertight capillary column nuts
Fingertight capillary column nuts
Finger-tight capillary column nut, for use with “compact” Agilent-style ferrules
Catalog # REST21311
Supplier:   Restek
Fingertight capillary column nuts
Catalog # REST21311
Supplier:  Restek
Fingertight capillary column nuts
Fingertight capillary column nuts
Finger-tight capillary column nut, for use with “compact” Agilent-style ferrules
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
These fingertight capillary column nuts rapidly tighten without wrenches and can be used with 0,25, 0,32, or 0,53 mm ID columns.
Specifications
  • Description:
    Finger-tight capillary column nut, for use with “compact” Agilent-style ferrules
  • Seq:
    01