Order Entry
Denmark
Orders LinkContactUsLinkComponent
Jumbo syringes

Jumbo syringes

1000 ml, 1000MAR-LL-GT
Catalog # REST21276
Supplier:   Restek
Jumbo syringes
Catalog # REST21276
Supplier:  Restek
Jumbo syringes
Jumbo syringes

1000 ml, 1000MAR-LL-GT

Price
9.020,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Clear acrylic syringes, ideal for holding and dispensing large volumes of gas.

  • Central port with a Luer lock fitting
  • Secondary port with septum nut
  • Detachable plunger stem
  • Easy sample storage

An adjustable plunger on the O-ring ensures that the syringe is gas-tight over a long period of time.

Specifications
  • Type:
    1000MAR-LL-GT
  • Volume:
    1000 ml
  • Seq:
    02
Accessories

Product Information

  • Catalog #
    For
    Availability
    Price
  • Syringe O-ring
    Syringe O-ring
    REST21278
    500 ml syringes
    Log in for availability
    1 * 1 Items

Product Information

  • Catalog #
    For
    Availability
    Price
  • Syringe O-ring
    Syringe O-ring
    REST21279
    1000 ml syringes
    Log in for availability
    1 * 1 Items

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.