AVFALLSLDA GLASAVFALL MED SÄCK 38L 1 * 25 items
SCAA95001039-1
: Essity
Frequently Bought Together
Pipette tips, Finntip™
: Thermo Fisher Scientific
VWR® Reagent Reservoirs
: VWR Collection
Ethanol absolute ≥99.5% USP, Ph. Eur.
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Nitrile gloves, Kimtech™ Blue Nitrile™
: KIMBERLY CLARK