Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Accessories for Storage Boxes

Partition with 11 slots, for A4 size, 60 mm high, 56 mm, 304 mm
Catalog # 216-0664
Supplier:   HOFSTATTER EBBESEN
Accessories for Storage Boxes
Catalog # 216-0664
Supplier:  HOFSTATTER EBBESEN
Accessories for Storage Boxes

Partition with 11 slots, for A4 size, 60 mm high, 56 mm, 304 mm

Price
55,10 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Partition with 11 slots, for A4 size, 60 mm high
  • Height:
    56 mm
  • Length:
    304 mm