Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for Storage Boxes

Accessories for Storage Boxes

Partition with 5 slots, for A4 and A5 size, 60 mm high, 56 mm, 218 mm
Catalog # 216-0658
Supplier:   HOFSTATTER EBBESEN
Accessories for Storage Boxes
Catalog # 216-0658
Supplier:  HOFSTATTER EBBESEN
Accessories for Storage Boxes

Partition with 5 slots, for A4 and A5 size, 60 mm high, 56 mm, 218 mm

Price
42,70 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Partition with 5 slots, for A4 and A5 size, 60 mm high
  • Height:
    56 mm
  • Length:
    218 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.