Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for Storage Boxes

Accessories for Storage Boxes

Lid, PS, clear, for A4 size, 12 mm, 315 mm
Catalog # 216-0680
Supplier:   HOFSTATTER EBBESEN
Accessories for Storage Boxes
Catalog # 216-0680
Supplier:  HOFSTATTER EBBESEN
Accessories for Storage Boxes

Lid, PS, clear, for A4 size, 12 mm, 315 mm

Price
297,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Lid, PS, clear, for A4 size
  • Height:
    12 mm
  • Length:
    315 mm