ROTILABO MAP SHEETS LENGTH 91 MM AND BROAD 59 MM 1 * 1 kg
Catalog # ROTH1656.1
Supplier: Roth Carl
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Beakers, Disposable
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Wipes and wipe rolls for general applications, WYPALL* L20 EXTRA / L20 EXTRA+
Supplier: KIMBERLY CLARK
Laboratory trays, low or high profile
Supplier: Gilac PROFESSIONNEL
OLAPARIB 1 * 1 g
Supplier: Thermo Fisher Scientific