Paraformaldehyde
Glass bottle for solids, 500 g
Katalog # ROTH0335.2
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Polyoxymethylene
- CAS nummer:30525-89-4
- Formel:(CH₂O)n
- MDL nummer:MFCD00133991
- Merck Index:13,07096
- Kogende Pt:135…136 °C (13 mmHg)
- Smeltende Pt:123 °C
- Tæthed:1.46 g/cm³ (20 °C)
- Flash Pt:70 °C
- Opbevaringstemperatur:Ambient
- FN:2213
- ADR:4.1,III
Specifikationer
Specifikationer
- Pack type:Glass bottle for solids
- Pk:500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



