Vancomycin hydrochloride
Glass bottle for solids, 1 g
Catalog # ROTH0242.3
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: (3S,6R,7R,22R,23S,26S,36R,38aR)-3-(2-Amino-2-oxoethyl)-44-[[2-O-(3-amino-2,3,6-trideoxy-3-C-methyl-α-L-lyxo-hexopyranosyl)-β-D-glucopyranosyl]oxy]-10,19-dichloro-2,3,4,5,6,7,23,24,25,26,36,37,38,38a-tetradecahydro-7,22,28,30,32-pentahydroxy-6-[[(2R)-4-methyl-2-(methylamino)-1-oxopentyl]amino]-2,5,24,38,39-pentaoxo-22H-8,11:18,21-Dietheno-23,36-(iminomethano)-13,16:31,35-dimetheno-1H,16H-[1,6,9]oxadiazacyclohexadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine-26-carboxylic acid xhydrochloride, Vancomycin supplement
- CAS Number:1404-93-9
- Formula:C₆₆H₇₅Cl₂N₉O₂₄·HCl
- MDL Number:MFCD03613611
- Molecular Weight:1485.71 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



