Starch (from wheat)
Glass bottle for solids, 1 kg
Catalog # ROTH9447.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:9005-25-8
- Formula:(C₆H₁₀O₅)n
- MDL Number:MFCD00082026
- Molecular Weight:162.14 g/mol
- Density:1.5 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:232-679-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



